Learn More
Sino Biological H5N1 NP Protein 100ug (Sino Biological)
Supplier:  Sino Biological 11958R34020
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Cambodia/NPH230032/2023) (H5N1)) nucleoprotein (EPI2419703) (Met1-Asn498) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-4262