Learn More
Sino Biological Influenza A H5N2 Hemagglutinin / HA Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11699RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H5N2 (A/American green-winge teal/California/HKWF609/07) HA / Hemagglutinin (Catalog#11699-V08H; ACF47563.1; Met 1-Gln 527). H5N2 (A/American green-winge teal/California/HKWF609/07) HA / Hemagglutinin specific IgG was purified by H5N2 (A/American green-winge teal/California/HKWF609/07) HA / Hemagglutinin affinity chromatography.
Catalog No. 50-241-0199