Learn More
Sino Biological Influenza A H5N2 Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11699RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H5N2 (A/American green-winge teal/California/HKWF609/07) HA / Hemagglutinin (Catalog#11699-V08H; ACF47563.1; Met 1-Gln 527). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8550