Learn More
Sino Biological Influenza A H5N8 Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11717RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H5N8 (A/duck/NY/191255-59/2002) HA / Hemagglutinin (Catalog#11717-V08H; AAP72011.1; Met 1-Gln 527). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8736