Learn More
Sino Biological H5N8 HA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11717RP0220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H5N8 (A/duck/NY/191255-59/2002) HA / Hemagglutinin (Catalog#11717-V08H; AAP72011.1; Met1-Gln527). H5N8 (A/duck/NY/191255-59/2002) HA / Hemagglutinin specific IgG was purified by H5N8 (A/duck/NY/191255-59/2002) HA / Hemagglutinin affinity chromatography.
Catalog No. 50-237-2512