Learn More
Sino Biological Influenza A H5N8 (A/Astrakhan/3212/2020) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 40932V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Astrakhan/3212/2020) (H5N8)) hemagglutinin (translated amino acids of EPI1846961) (Met1-Arg345), termed as HA1, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-240-8587