Learn More
Alkali Scientific H6002 Heated Incu-Mixer MP2 Vo
SDP
Catalog No. 503029628
Encompass_Preferred
The new Incu-Mixer MP series is designed to provide an optimal method for vortexing and incubating microplates Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample Additionally the heated lid minimizes evaporation and prevents condensation A compact 2mm orbit ensures thorough mixing in microplates without splashing Speed time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy Designed for application flexibility the Incu-Mixer series is available in two and four-plate capacities both capable of accepting all standard plates up to 40mm high Independent time temperature and speed controls allow the mixer to function as 3 machines in one a multi-plate incubator vortexer or a combination of the two The Incu-Mixer MP series is ideal for a wide variety of molecular biology applications including immunochemical reactions e
Catalog No. 50-302-9628 Supplier Alkali Scientific Supplier No. RS7507
The new Incu-Mixer MP series is designed to provide an optimal method for vortexing and incubating microplates Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample Additionally the heated lid minimizes evaporation and prevents condensation A compact 2mm orbit ensures thorough mixing in microplates without splashing Speed time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy Designed for application flexibility the Incu-Mixer series is available in two and four-plate capacities both capable of accepting all standard plates up to 40mm high Independent time temperature and speed controls allow the mixer to function as 3 machines in one a multi-plate incubator vortexer or a combination of the two The Incu-Mixer MP series is ideal for a wide variety of molecular biology applications including immunochemical reactions e