Learn More
Alkali Scientific H6004 Heated Incu-Mixer MP2 Vo
SDP
Catalog No. 503029555
Encompass_Preferred
The new H6004 Incu-Mixer by Alkali Scientificis designed to provide an optimal method for vortexing and incubating microplates Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample Additionally the heated lid minimizes evaporation and prevents condensation A compact 2mm orbit ensures thorough mixing in microplates without splashing Speed time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy Designed for application flexibility the Incu-Mixer series is available in two and four-plate capacities The smaller unit accommodates all standard plates up to 20mm high while the larger unit will support plates up to 46mm Independent time temperature and speed controls allow the mixer to function as 3 machines in one a multi-plate incubator vortexer or a combination of the two The Incu-Mixer H6004 MP is ideal for a wide variety
Catalog No. 50-302-9555 Supplier Alkali Scientific Supplier No. RS7508
The new H6004 Incu-Mixer by Alkali Scientificis designed to provide an optimal method for vortexing and incubating microplates Simultaneous heating from both the bottom surface and the lid provide temperature uniformity throughout the sample Additionally the heated lid minimizes evaporation and prevents condensation A compact 2mm orbit ensures thorough mixing in microplates without splashing Speed time and temperature are all displayed in real time on the large LCD screen while an internal validation system constantly monitors all parameters for accuracy Designed for application flexibility the Incu-Mixer series is available in two and four-plate capacities The smaller unit accommodates all standard plates up to 20mm high while the larger unit will support plates up to 46mm Independent time temperature and speed controls allow the mixer to function as 3 machines in one a multi-plate incubator vortexer or a combination of the two The Incu-Mixer H6004 MP is ideal for a wide variety