Learn More
MSC Value Collection Halogen Lamp, 12 Volt, Rectangular Base, 1 ea
Catalog No. 502868796
Encompass
Halogen bulbs produce more bright light (Lumens) for less watts compared to standard incandescent bulbs. They also produce the most heat. Halogen bulbs are an economical solution that provides a bright, white light with excellent beam qualities for various applications.
  • Produce more bright light (lumens) for less watts compared to standard incandescent bulbs
  • Produce the most heat
  • Economical solution that provides a bright, white light with excellent beam qualities
Catalog No. 50-286-8796 Supplier MSC Supplier No. 66451683
Halogen bulbs produce more bright light (Lumens) for less watts compared to standard incandescent bulbs. They also produce the most heat. Halogen bulbs are an economical solution that provides a bright, white light with excellent beam qualities for various applications.
  • Produce more bright light (lumens) for less watts compared to standard incandescent bulbs
  • Produce the most heat
  • Economical solution that provides a bright, white light with excellent beam qualities