Learn More
Grainger Schneider Electric Pushbutton Head, Flush, Blue, 22 mm
Catalog No. 502759997
Encompass_Preferred
The Harmony XB4 series flush blue pushbutton head provides a durable, ergonomic interface for industrial machinery control. Designed for a standard 22 mm panel cutout, this non-illuminated operator features a spring return impulse mechanism and a protective clear boot, making it suitable for high-hygiene environments by preventing bacterial growth in gaps. The unit features a robust chromium-plated metal bezel and is built to withstand demanding conditions.
  • IP66 rating for dust and water resistance
  • Impact and vibration resistant design for long-term reliability
  • Snap-fit assembly for simple installation and replacement
  • Operating temperature range of -40°C to 70°C
  • Clear boot construction prevents bacteria accumulation
  • Compatible with standard 22 mm panel mount configurations
Catalog No. 50-275-9997 Supplier Grainger Supplier No. 15V820
The Harmony XB4 series flush blue pushbutton head provides a durable, ergonomic interface for industrial machinery control. Designed for a standard 22 mm panel cutout, this non-illuminated operator features a spring return impulse mechanism and a protective clear boot, making it suitable for high-hygiene environments by preventing bacterial growth in gaps. The unit features a robust chromium-plated metal bezel and is built to withstand demanding conditions.
  • IP66 rating for dust and water resistance
  • Impact and vibration resistant design for long-term reliability
  • Snap-fit assembly for simple installation and replacement
  • Operating temperature range of -40°C to 70°C
  • Clear boot construction prevents bacteria accumulation
  • Compatible with standard 22 mm panel mount configurations