Learn More
ATCC H69AR; Lung Carcinoma; Human (Homo sapiens)
Catalog No. 502383376
Encompass
H69AR is an epithelial cell isolated from the lung of a 55-year-old White male patient with carcinoma. This cell line was deposited by S. Cole and can be used for cancer research.
Catalog No. 50-238-3376 Supplier ATCC Supplier No. CRL11351
H69AR is an epithelial cell isolated from the lung of a 55-year-old White male patient with carcinoma. This cell line was deposited by S. Cole and can be used for cancer research.