Learn More
Sino Biological Influenza A H6N1 (A/northern shoveler/California/HKWF115/2007) Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11723RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H6N1 (A/northern shoveler/California/HKWF115/2007) Hemagglutinin / HA (Catalog#11723-V08H; ACE81692.1; Met1-Gln529). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8015