Learn More
Aviva Systems Biology Corp H6PD Antibody - N-terminal region (ARP51206_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51206_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against H6PD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human H6PD. Alternate names: DKFZp686A01246, G6PDH, GDH, MGC87643ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60515