Learn More
Biotang Inc Influenza A H7N7 (A/chicken/Netherlands/1/03) Hemagglutinin Protein (HA1 Subunit) (His Tag), 10ug, an N-terminal segment (Met 1-Arg 344) of Influenza A virus (A/chicken/Netherlands/1/03 (H7N7)) hemagglutinin (AAR02639.1), termed as HA1
Supplier:  Biotang Inc RPR9458
Encompass_Preferred
Influenza A H7N7 (A/chicken/Netherlands/1/03) Hemagglutinin Protein (HA1 Subunit) (His Tag), 10ug, an N-terminal segment (Met 1-Arg 344) of Influenza A virus (A/chicken/Netherlands/1/03 (H7N7)) hemagglutinin (AAR02639.1), termed as HA1
Catalog No. 50-751-5082