Learn More
Biotang Inc Influenza A H7N7 (A/chicken/Netherlands/1/03) Hemagglutinin / HA Protein (His Tag), 10ug, the extracellular domain of Influenza A virus (A/chicken/Netherlands/1/03 (H7N7)) (AAR02639.1) hemagglutinin (Met 8-Phe 544) (Native, HA1+HA2).
Supplier:  Biotang Inc RPR9518
Encompass_Preferred
Influenza A H7N7 (A/chicken/Netherlands/1/03) Hemagglutinin / HA Protein (His Tag), 10ug, the extracellular domain of Influenza A virus (A/chicken/Netherlands/1/03 (H7N7)) (AAR02639.1) hemagglutinin (Met 8-Phe 544) (Native, HA1+HA2).
Catalog No. 50-751-5081