Learn More
Sino Biological H7N7 HA PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50468M02H1
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H7N7 (A/Netherlands/219/2003) Hemagglutinin / HA ( Catalog#11082-V08B; AAR02640.1; Met8-Phe544). Hemagglutinin/HA specific IgG was purified by Influenza A H7N7 (A/Netherlands/219/2003) Hemagglutinin / HA affinity chromatography.
Catalog No. 50-237-4499