Learn More
Sino Biological Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin / HA Protein (His Tag), Biotinylated
Catalog No. 502993465
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Shanghai/1/2013(H7N9)) hemagglutinin (translated amino acid of EPI744956) (Met1-Val524) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-299-3465 Supplier Sino Biological Supplier No. 40104V08HB20
A DNA sequence encoding the Influenza A virus (A/Shanghai/1/2013(H7N9)) hemagglutinin (translated amino acid of EPI744956) (Met1-Val524) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.