Learn More
Sino Biological H7N9 HA PAb 20ul (Sino Biological)
Supplier:  Sino Biological 40104T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H7N9 (A/Shanghai/1/2013) HA (Catalog#40104-V08B; Met1-Val524). HA specific IgG was purified by Influenza A H7N9 (A/Shanghai/1/2013) HA affinity chromatography.
Catalog No. 50-237-3191