Learn More
Sino Biological H7N9 HA PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14905MM07100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin / HA (Catalog#40103-V08B; EPI439507; Met1-Val524). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-4445