Learn More
Sino Biological Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 40104RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin / HA (Catalog#40104-V08B; Met1-Val524). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8086