Learn More
Cayman Chemical H-8 (hydrochloride), Purity: ≥98%, CAS: 113276-94-1, 10 mg
SDP
Catalog No. 502887966
Encompass_Preferred
A potent inhibitor of PKA and PKG that shows moderate inhibition for PKC and MLCK with Ki values of 1.2, 0.48, 15, and 68 µM, respectively
Catalog No. 50-288-7966 Supplier Cayman Chemical Supplier No. 1001024910 MG
A potent inhibitor of PKA and PKG that shows moderate inhibition for PKC and MLCK with Ki values of 1.2, 0.48, 15, and 68 µM, respectively