Learn More
Cayman Chemical H-89 (hydrochloride), Purity: ≥98%, CAS: 130964-39-5, 25 mg
SDP
Catalog No. 502888596
Encompass_Preferred
A potent, non-selective inhibitor of PKA with an IC50 value of 0.14 µM (Ki = 48 nM) that is widely used to disrupt PKA signaling; inhibits S6K1, MSK1, ROCK2, PKBa, and MAPKAP-K1b with IC50 values of 0.08, 0.12, 0.27, 2.6, and 2.8 µM, respectively
Catalog No. 50-288-8596 Supplier Cayman Chemical Supplier No. 1001055625 MG
A potent, non-selective inhibitor of PKA with an IC50 value of 0.14 µM (Ki = 48 nM) that is widely used to disrupt PKA signaling; inhibits S6K1, MSK1, ROCK2, PKBa, and MAPKAP-K1b with IC50 values of 0.08, 0.12, 0.27, 2.6, and 2.8 µM, respectively