Learn More
Sino Biological Influenza A H8N4 Hemagglutinin / HA Antibody, Mouse MAb
Supplier:  Sino Biological 11722MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A virus H8N4 Hemagglutinin (rInfluenza A virus H8N4 Hemagglutinin; Met 1-Lys 529; ABB87729.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8565