Learn More
Sino Biological Influenza A H8N4 Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11722RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A virus H8N4 Hemagglutinin (Catalog#11722-V08H; Met 1-Lys 529; ABB87729.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-7775