Learn More
Sino Biological Influenza A H9N2 Hemagglutinin / HA Antibody (HRP), Rabbit MAb
Supplier:  Sino Biological 11229R106H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A virus H9N2 Hemagglutinin, and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-4771