Learn More
Sino Biological Influenza A H9N2 (A/Hong Kong/1073/99) Hemagglutinin / HA Antibody, Mouse MAb
Supplier:  Sino Biological 11229MM13100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H9N2 (A/Hong Kong/1073/99) Hemagglutinin / HA ( Catalog#11229-V08H; NP_859037.1; Met1-Lys523). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4773