Learn More
Sino Biological Influenza A H9N2 Hemagglutinin / HA Antibody (Biotin), Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11229RP02B20
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A virus H9N2 HA / Hemagglutinin. HA specific IgG was purified by H9N2 HA / Hemagglutinin affinity chromatography and then biotinylated.
Catalog No. 50-255-4768