Learn More
Sino Biological Influenza H9N2 (A/chicken/Hong Kong/G9/1997) Hemagglutinin / HA1 Antibody, Rabbit PAb
Supplier:  Sino Biological 40036RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza H9N2 (A/chicken/Hong Kong/G9/1997) Hemagglutinin / HA1 ( Catalog#40036-V08H1; AAF00701.1; Met1-Arg338). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8948