Learn More
Sino Biological Influenza A H9N2 (A/Hong Kong/1073/99) Neuraminidase / NA Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40040T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H9N2 (A/Hong Kong/1073/99) Neuraminidase / NA ( Catalog#40040-V07H; NP_859038.1; His36-Ile469). Influenza A H9N2 (A/Hong Kong/1073/99) Neuraminidase / NA specific IgG was purified by Influenza A H9N2 (A/Hong Kong/1073/99) Neuraminidase / NA affinity chromatography.
Catalog No. 50-237-4208