Learn More
Medchemexpress LLC HY-100984 5mg Medchemexpress, HA-100 CAS:84468-24-6 Purity:>98%
SDP
Catalog No. 501932005
Encompass_Preferred
Medchemexpress, HY-100984 5mg HA-100 CAS:84468-24-6 HA-100 is an inhibitor of cGMP-dependent protein kinase ( PKG ), cAMP-dependent protein kinase ( PKA ), protein kinase C ( PKC ) and MLC-kinase with IC 50 s of 4, 8, 12 and 240 μM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2005 Supplier Medchemexpress LLC Supplier No. HY1009845MG
Medchemexpress, HY-100984 5mg HA-100 CAS:84468-24-6 HA-100 is an inhibitor of cGMP-dependent protein kinase ( PKG ), cAMP-dependent protein kinase ( PKA ), protein kinase C ( PKC ) and MLC-kinase with IC 50 s of 4, 8, 12 and 240 μM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.