Learn More
Selleck Chemical LLC HA-100 dihydrochloride S2964-5mg
SDP
Catalog No. 503112821
Encompass_Preferred
HA-100 dihydrochloride is an inhibitor of cGMP-dependent protein kinase (PKG) cAMP-dependent protein kinase (PKA) protein kinase C (PKC) and MLC-kinase with IC50 of 4 M 8 M 12 M and 240 M respectively
Catalog No. 50-311-2821 Supplier Selleck Chemical LLC Supplier No. S29645MG
HA-100 dihydrochloride is an inhibitor of cGMP-dependent protein kinase (PKG) cAMP-dependent protein kinase (PKA) protein kinase C (PKC) and MLC-kinase with IC50 of 4 M 8 M 12 M and 240 M respectively