Learn More
Enzo Life Sciences HA-100 . dihydrochloride (5mg). CAS: 84468-24-6
SDP
Supplier:  Enzo Life Sciences ALX270069M005
Encompass_Preferred
Inhibitor of myosin light chain kinase, cAMP-dependent protein kinase (PKA) and protein kinase C (PKC). Alternative name: 1-(5-Isoquinolinylsulfonyl)piperazine . 2HCl. Formula: C13H15N3O2S . 2HCl. MW: 350.3. Purity: ≥98% (HPLC). Solubility: Soluble in water or DMSO. Long Term Storage: +4°C.
Catalog No. 50-200-8397