Learn More
STEMCELL Technologies HA-100 (Dihydrochloride), Size: 5 mg
SDP
Catalog No. 501975928
Encompass
HA-100 is an isoquinoline compound with an added piperazinylsulfonyl group that acts as an inhibitor of protein kinases (PKs), including PKA, PKC, and PKG (IC₅₀ = 8, 12, and 4 µM, respectively) (Hagiwara et al.).REPROGRAMMING· Increases human fibroblast reprogramming efficiency with PD0325901, CHIR99021, A83-01 and hLIF (Yu et al.).MAINTENANCE AND SELF-RENEWAL· Improves single cell survival and supports high cloning efficiency in human pluripotent stem cell cultures (Chen et al.).
Catalog No. 50-197-5928 Supplier STEMCELL Technologies Supplier No. 72482
HA-100 is an isoquinoline compound with an added piperazinylsulfonyl group that acts as an inhibitor of protein kinases (PKs), including PKA, PKC, and PKG (IC₅₀ = 8, 12, and 4 µM, respectively) (Hagiwara et al.).REPROGRAMMING· Increases human fibroblast reprogramming efficiency with PD0325901, CHIR99021, A83-01 and hLIF (Yu et al.).MAINTENANCE AND SELF-RENEWAL· Improves single cell survival and supports high cloning efficiency in human pluripotent stem cell cultures (Chen et al.).