Learn More
TARGETMOL CHEMICALS INC HA-1004 5MG
SDP
Catalog No. 502580603
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. HA-1004 is a inhibitor of PKG PKA PKC and MLC. purity: 99%
Catalog No. 50-258-0603 Supplier TARGETMOL CHEMICALS INC Supplier No. T275255MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. HA-1004 is a inhibitor of PKG PKA PKC and MLC. purity: 99%