Learn More
Cayman Chemical HA-100 (hydrochloride), Purity: ≥95%, CAS: 210297-47-5, 50 mg
SDP
Catalog No. 502970919
Encompass_Preferred
An inhibitor of PKA, PKC, and PKG (IC50s = 8, 12, and 4 µM, respectively); less effectively blocks the activity of MLCK (IC50 = 240 µM)
Catalog No. 50-297-0919 Supplier Cayman Chemical Supplier No. 1495550 MG
An inhibitor of PKA, PKC, and PKG (IC50s = 8, 12, and 4 µM, respectively); less effectively blocks the activity of MLCK (IC50 = 240 µM)