Learn More
Enzo Life Sciences HA.11 monoclonal antibody (16B12) (purified) (1ml)
SDP
Supplier:  Enzo Life Sciences ENZABS1201000
Encompass_Preferred
Clone: 16B12. Isotype: IgG1. Immunogen: Twelve amino acid peptide CYPYDVPDYASL. Specificity: Recognizes the influenza hemagglutinin epitope (YPYDVPDYA) which has been used extensively as a general epitope tag in expression vectors. Recognizes HA epitopes located in the middle of protein sequences as well as at the N- or C-terminus. Applications: IF, IHC, IP, WB. Formulation: Liquid. In PBS containing 50% glycerol and 0.03% thimerosal.
Catalog No. 50-201-2682