Learn More
Enzo Life Sciences HA.11 polyclonal antibody (purified) (200µl)
SDP
Catalog No. 502012752
Encompass_Preferred
Immunogen: Twelve amino acid peptide CYPYDVPDYASL. Specificity: Recognizes the influenza hemagglutinin epitope (YPYDVPDYA). Applications: IP, WB. Formulation: Liquid. In PBS containing 0.03% thimerosal.
Catalog No. 50-201-2752 Supplier Enzo Life Sciences Supplier No. ENZABS2030200
Immunogen: Twelve amino acid peptide CYPYDVPDYASL. Specificity: Recognizes the influenza hemagglutinin epitope (YPYDVPDYA). Applications: IP, WB. Formulation: Liquid. In PBS containing 0.03% thimerosal.