Learn More
Cayman Chemical HA-1100hydrochlorIde 1mg
SDP
Catalog No. 502969552
Encompass_Preferred
A cell-permeable inhibitor of ROCK (Ki = 150 nM); less effectively inhibits PKA (Ki = 2.2 μM)
Catalog No. 50-296-9552 Supplier Cayman Chemical Supplier No. 144131 MG
A cell-permeable inhibitor of ROCK (Ki = 150 nM); less effectively inhibits PKA (Ki = 2.2 μM)