Learn More
TA Instruments HA Aluminum, 20 mm Plate, Upper Plate for Use with Lower Peltier Plate, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580774 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, HA Aluminum, 20 mm Plate is an upper plate for use with the lower Peltier plate. It has Smart Swap 2 geometry and is for use with HR X0 instruments only.
Catalog No. 50-005-80774 Supplier TA Instruments Supplier No. 513200.945
Waters TA Instruments HR X0, HA Aluminum, 20 mm Plate is an upper plate for use with the lower Peltier plate. It has Smart Swap 2 geometry and is for use with HR X0 instruments only.