Learn More
TA Instruments HA Aluminum, Double Gap Rotor, Inner Radius 16 mm, Outer Radius 17.5 mm, Height 53 mm, HR X0
Catalog No. 5000580761 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, HA Aluminum, Double Gap Rotor has an inner radius of 16 mm, outer radius of 17.5 mm, and height of 53 mm.
Catalog No. 50-005-80761 Supplier TA Instruments Supplier No. 546049.941
Waters TA Instruments HR X0, HA Aluminum, Double Gap Rotor has an inner radius of 16 mm, outer radius of 17.5 mm, and height of 53 mm.