Learn More
TA Instruments HA Aluminum Grooved Concentric Cylinder Cup, Stops Sample Slipping, for Large or Small Diameter Vaned Rotors, Inner/Outer Diameter 30/34 mm, Groove Depth 4 mm, for HR X0, DHR and AR-Series Peltier Jacket
Catalog No. 5000580641 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HA Aluminum Grooved Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket systems. The groove stops sample slipping at the cup wall. It is designed for use with large or small diameter vaned rotors. The inner diameter is 30 mm, the outer diameter is 34 mm, and the groove depth is 4 mm. Sample volume depends on rotor selection.
Catalog No. 50-005-80641 Supplier TA Instruments Supplier No. 545610.901
Waters TA Instruments HA Aluminum Grooved Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket systems. The groove stops sample slipping at the cup wall. It is designed for use with large or small diameter vaned rotors. The inner diameter is 30 mm, the outer diameter is 34 mm, and the groove depth is 4 mm. Sample volume depends on rotor selection.