Learn More
TA Instruments HA Aluminum Large Diameter Concentric Cylinder Cup, Radius 22 mm, Removable Stainless Steel Base, Compatible with All Upper Rotors, Sample Volume Depends on Rotor Selection, for HR X0, DHR and AR-Series Peltier Jacket
Catalog No. 5000580860 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HA Aluminum Large Diameter Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket. It has a removable stainless steel base, a radius of 22 mm, and is compatible with all upper rotors. Sample volume depends on rotor selection.
Catalog No. 50-005-80860 Supplier TA Instruments Supplier No. 545615.901
Waters TA Instruments HA Aluminum Large Diameter Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket. It has a removable stainless steel base, a radius of 22 mm, and is compatible with all upper rotors. Sample volume depends on rotor selection.