Learn More
TA Instruments HA Aluminum Standard Concentric Cylinder Cup, Compatible with Upper Conical DIN, Recessed End, Vane, Wide Gap Vane Rotors, Sample Volume Depends on Rotor Selection, for HR X0, DHR and AR-Series Peltier Jacket
Catalog No. 5000580661 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HA Aluminum Standard Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket. It is compatible with upper conical DIN, recessed end, vane, and wide gap vane rotors. Sample volume depends on rotor selection.
Catalog No. 50-005-80661 Supplier TA Instruments Supplier No. 545700.901
Waters TA Instruments HA Aluminum Standard Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket. It is compatible with upper conical DIN, recessed end, vane, and wide gap vane rotors. Sample volume depends on rotor selection.