Learn More
TA Instruments HA Aluminum Starch Rotor Concentric Cylinder Cup, Use with Upper Starch Pasting Rotor P/N 545712.941/545712.901/545751.001 or Conical Rotor P/N 545713.941/545713.901/545761.001, for HR X0, DHR and AR-Series Peltier Jacket
Catalog No. 5000580700 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HA Aluminum Starch Rotor Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket systems. It is designed for use with the upper starch pasting rotor (P/N 545712.941 for HR X0, 545712.901 for DHR, 545751.001 for AR-Series) or the conical rotor (P/N 545713.941 for HR X0, 545713.901 for DHR, 545761.001 for AR-Series).
Catalog No. 50-005-80700 Supplier TA Instruments Supplier No. 545666.901
Waters TA Instruments HA Aluminum Starch Rotor Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket systems. It is designed for use with the upper starch pasting rotor (P/N 545712.941 for HR X0, 545712.901 for DHR, 545751.001 for AR-Series) or the conical rotor (P/N 545713.941 for HR X0, 545713.901 for DHR, 545761.001 for AR-Series).