Learn More
Medchemexpress LLC HA/Hemagglutinin H1 100ug | 100ug
SDP
Catalog No. 5000186229
Encompass_Preferred
The hemagglutinin HA1 protein is essential for attachment of virions to host cells by binding to sialic acid-containing receptors inducing virion internalization via clathrin-dependent or clathrin- and caveolin-independent pathways As a class I viral fusion protein HA1 determines host range restriction and virulence mediating fusion between the virion membrane and the endosomal membrane Hemagglutinin/HA Protein H12N1 (ABB88099 sf9 His) is the recombinant Virus-derived Hemagglutinin/HA protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-86229 Supplier Medchemexpress LLC Supplier No. HYP77019100UG
The hemagglutinin HA1 protein is essential for attachment of virions to host cells by binding to sialic acid-containing receptors inducing virion internalization via clathrin-dependent or clathrin- and caveolin-independent pathways As a class I viral fusion protein HA1 determines host range restriction and virulence mediating fusion between the virion membrane and the endosomal membrane Hemagglutinin/HA Protein H12N1 (ABB88099 sf9 His) is the recombinant Virus-derived Hemagglutinin/HA protein expressed by Sf9 insect cells with C-His labeled tag