Learn More
Medchemexpress LLC HA/Hemagglutinin H1 100ug
SDP
Supplier:  Medchemexpress LLC HYP74039100UG
Encompass_Preferred
The hemagglutinin (HA) protein attaches viral particles to host cells by binding to sialic acid-containing receptors and induces viral particle internalization through clathrin-dependent endocytosis HA also promotes an alternative clathrin- and caveolin-independent pathway for some virions HA/Hemagglutinin Protein H1N1 (Biotinylated ACP41105 sf9 His) is the recombinant Virus-derived HA/Hemagglutinin protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-79932