Learn More
Medchemexpress LLC HA/Hemagglutinin H1 10ug | 10ug
SDP
Supplier:  Medchemexpress LLC HYP7702010UG
Encompass_Preferred
The HA1/Hemagglutinin protein binds cell surface receptors facilitating virus attachment It determines host range and virulence mediating fusion of viral and endosomal membranes Acidic endosomes trigger HA2 conformational change releasing fusion peptide Multiple HA1 trimers form fusion pore Hemagglutinin/HA Protein H14N5 (P26136 sf9 His) is the recombinant Virus-derived Hemagglutinin/HA protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-85952