Learn More
Medchemexpress LLC HA/Hemagglutinin H1 10ug
Supplier: Medchemexpress LLC HYP7403210UG
The hemagglutinin (HA) protein attaches viral particles to host cells by binding to sialic acid-containing receptors and induces viral particle internalization through clathrin-dependent endocytosis HA also promotes an alternative clathrin- and caveolin-independent pathway for some virions HA/Hemagglutinin Protein H1N1 (QEM24663 sf9 His) is the recombinant Virus-derived HA/Hemagglutinin protein expressed by Sf9 insect cells with C-His labeled tag