Learn More
Medchemexpress LLC HA/Hemagglutinin H1 10ug
SDP
Catalog No. 5000179459
Encompass_Preferred
The hemagglutinin (HA) protein attaches viral particles to host cells by binding to sialic acid-containing receptors and induces viral particle internalization through clathrin-dependent endocytosis HA also promotes an alternative clathrin- and caveolin-independent pathway for some virions HA/Hemagglutinin Protein H1N1 (AAD17229 HEK293 His) is the recombinant Virus-derived HA/Hemagglutinin protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79459 Supplier Medchemexpress LLC Supplier No. HYP7409210UG
The hemagglutinin (HA) protein attaches viral particles to host cells by binding to sialic acid-containing receptors and induces viral particle internalization through clathrin-dependent endocytosis HA also promotes an alternative clathrin- and caveolin-independent pathway for some virions HA/Hemagglutinin Protein H1N1 (AAD17229 HEK293 His) is the recombinant Virus-derived HA/Hemagglutinin protein expressed by HEK293 with C-His labeled tag