Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HA/hemagglutinin protein, H1N1 (EPI2397519, Sf9, His) | >95.0% | 10UG
SDP

Catalog No. 5000174026
Encompass_Preferred

HA/Hemagglutinin Protein, H1N1 (EPI2397519, sf9, His) is a recombinant influenza A surface glycoprotein produced in Sf9 insect cells with a C-terminal His tag. Supplied as a filtered, buffered solution for research use, the protein is characterized by high purity and a defined molecular weight suitable for biochemical and immunological assays.

  • Recombinant HA from H1N1 influenza, sequence accession EPI2397519.
  • Expressed in Sf9 insect cells for eukaryotic post-translational modifications.
  • C-terminal His tag for purification and detection.
  • Supplied in 20 mM Tris, 300 mM NaCl, 10% glycerol, pH 8.0, 0.2 μm filtered.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 65.49 kDa under reducing conditions.
  • For research use only; store at -80°C and avoid repeated freeze-thaw cycles.

Catalog No. 50-001-74026 Supplier Medchemexpress LLC Supplier No. HYP70411110UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HA/Hemagglutinin Protein, H1N1 (EPI2397519, sf9, His) is a recombinant influenza A surface glycoprotein produced in Sf9 insect cells with a C-terminal His tag. Supplied as a filtered, buffered solution for research use, the protein is characterized by high purity and a defined molecular weight suitable for biochemical and immunological assays.

  • Recombinant HA from H1N1 influenza, sequence accession EPI2397519.
  • Expressed in Sf9 insect cells for eukaryotic post-translational modifications.
  • C-terminal His tag for purification and detection.
  • Supplied in 20 mM Tris, 300 mM NaCl, 10% glycerol, pH 8.0, 0.2 μm filtered.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 65.49 kDa under reducing conditions.
  • For research use only; store at -80°C and avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.